Gene Transcripts

Gene Symbol
Phip
Entrez Gene ID
315843
Gene Name
pleckstrin homology domain interacting protein
Gene Aliases
RGD1564964
Chromosome Location
Chr. 8: 90387339 - 90483964 on Build Rnor_6.0
UniGene ID
Rn.232332
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006243470.1 XP_006243532.1 37 5280

View Full Product Details