Gene Transcripts

Gene Symbol
Nrk
Entrez Gene ID
315907
Gene Name
Nik related kinase
Gene Aliases
RGD1566163
Chromosome Location
Chr. X: 109940439 - 110036361 on Build Rnor_6.0
UniGene ID
Rn.214057
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191797.1 NP_001178726.1 58

View Full Product Details