Gene Transcripts

Gene Symbol
March1
Entrez Gene ID
361135
Gene Name
membrane-associated ring finger (C3HC4) 1, E3 ubiquitin protein ligase
Gene Aliases
RGD1305148
Chromosome Location
Chr. 16: 24998709 - 25524786 on Build Rnor_6.0
UniGene ID
Rn.225169
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135838.1 NP_001129310.1 6 633
XM_008771133.1 XP_008769355.1 4 771
XM_008771134.1 XP_008769356.1 4 840

Gene Symbol
LOC100910061
Entrez Gene ID
100910061
Gene Name
E3 ubiquitin-protein ligase MARCH1-like
Gene Aliases
Chromosome Location
Chr. 16: 26080974 - 26098930 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006253055.2 XP_006253117.1 2 746

View Full Product Details