Gene Transcripts

Gene Symbol
Akr1cl
Entrez Gene ID
361267
Gene Name
aldo-keto reductase family 1, member C-like
Gene Aliases
Akr1cl1
Chromosome Location
Chr. 17: 69835338 - 69862093 on Build Rnor_6.0
UniGene ID
Rn.206135
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109900.1 NP_001103370.1 3 380

View Full Product Details