Gene Transcripts

Gene Symbol
Mocos
Entrez Gene ID
361300
Gene Name
molybdenum cofactor sulfurase
Gene Aliases
Chromosome Location
Chr. 18: 16590100 - 16636084 on Build Rnor_6.0
UniGene ID
Rn.51875
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108425.1 NP_001101895.1 11 2344
XM_006254466.2 XP_006254528.1 10 2511

View Full Product Details