Gene Transcripts

Gene Symbol
Clca4
Entrez Gene ID
362053
Gene Name
chloride channel accessory 4
Gene Aliases
Clca6, Prp3
Chromosome Location
Chr. 2: 250843165 - 250864060 on Build Rnor_6.0
UniGene ID
Rn.44141
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201419.1 NP_958822.1 13 2072
GenBank mRNA AY333961.1 - 13 2072

View Full Product Details