Gene Transcripts

Gene Symbol
Ube3c
Entrez Gene ID
362294
Gene Name
ubiquitin protein ligase E3C
Gene Aliases
RGD1559986
Chromosome Location
Chr. 4: 2505843 - 2606711 on Build Rnor_6.0
UniGene ID
Rn.9242
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762667.1 XP_008760889.1 23 3569
GenBank mRNA FQ214186.1 - 2 179

View Full Product Details