Gene Transcripts

Gene Symbol
Plekhm2
Entrez Gene ID
313667
Gene Name
pleckstrin homology domain containing, family M (with RUN domain) member 2
Gene Aliases
Chromosome Location
Chr. 5: 160182556 - 160219770 on Build Rnor_6.0
UniGene ID
Rn.144389
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191767.1 NP_001178696.1 4 358

View Full Product Details