Gene Transcripts

Gene Symbol
Tubgcp6
Entrez Gene ID
362980
Gene Name
tubulin, gamma complex associated protein 6
Gene Aliases
Chromosome Location
Chr. 7: 130080895 - 130102264 on Build Rnor_6.0
UniGene ID
Rn.24182
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108748.1 NP_001102218.1 10 1939
XM_006242217.2 XP_006242279.1 10 2345

View Full Product Details