Gene Transcripts

Gene Symbol
Srp68
Entrez Gene ID
363707
Gene Name
signal recognition particle 68
Gene Aliases
Chromosome Location
Chr. 10: 105123324 - 105150080 on Build Rnor_6.0
UniGene ID
Rn.68
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108840.1 NP_001102310.1 1772
XM_008768381.1 XP_008766603.1 9 2486
GenBank mRNA BC166787.1 - 1669

View Full Product Details