Gene Transcripts

Gene Symbol
Epg5
Entrez Gene ID
364902
Gene Name
ectopic P-granules autophagy protein 5 homolog (C. elegans)
Gene Aliases
RGD1306286
Chromosome Location
Chr. 18: 74299966 - 74397182 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772212.1 XP_008770434.1 15, 16 2935

View Full Product Details