Gene Transcripts

Gene Symbol
Bank1
Entrez Gene ID
365948
Gene Name
B-cell scaffold protein with ankyrin repeats 1
Gene Aliases
LRRGT00104
Chromosome Location
Chr. 2: 241273763 - 241545250 on Build Rnor_6.0
UniGene ID
Rn.33991
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047918.2 NP_001041383.2 12 2022
GenBank mRNA FQ231577.1 - 11 1702

View Full Product Details