Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001270586.1 | NP_001257515.1 | 13, 14 | 1748 | |
NM_001270587.1 | NP_001257516.1 | 12, 13 | 1754 | ||
NM_031650.3 | NP_113838.1 | 13, 14 | 1853 | ||
GenBank mRNA | AF147740.1 | - | 12, 13 | 1634 | |
AF208545.2 | - | 13, 14 | 1853 | ||
AF217450.2 | - | 12, 13 | 1754 | ||
AJ271682.1 | - | 13, 14 | 1854 |