Gene Transcripts

Gene Symbol
Hmox1
Entrez Gene ID
24451
Gene Name
heme oxygenase (decycling) 1
Gene Aliases
HEOXG, Heox, Hmox, Ho-1, Ho1, hsp32
Chromosome Location
Chr. 19: 14508634 - 14515455 on Build Rnor_6.0
UniGene ID
Rn.3160
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012580.2 NP_036712.1 3 324
GenBank mRNA BC091164.1 - 3 324
FQ228696.1 - 2 198

View Full Product Details