Gene Transcripts

Gene Symbol
Map3k19
Entrez Gene ID
289001
Gene Name
mitogen-activated protein kinase kinase kinase 19
Gene Aliases
RGD1566400, Ysk4
Chromosome Location
Chr. 13: 44524067 - 44552748 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002727999.3 XP_002728045.3 10 4338

View Full Product Details