Gene Transcripts

Gene Symbol
Ube2s
Entrez Gene ID
292588
Gene Name
ubiquitin-conjugating enzyme E2S
Gene Aliases
RGD1564746
Chromosome Location
Chr. 1: 72580424 - 72584484 on Build Rnor_6.0
UniGene ID
Rn.120398
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106224.2 NP_001099694.1 4 489
GenBank mRNA BC169077.1 - 4 489

View Full Product Details