Gene Transcripts

Gene Symbol
Arrdc2
Entrez Gene ID
306344
Gene Name
arrestin domain containing 2
Gene Aliases
ILAD1
Chromosome Location
Chr. 16: 20352480 - 20356612 on Build Rnor_6.0
UniGene ID
Rn.22831
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107303.1 NP_001100773.1 4, 5 942

View Full Product Details