Gene Transcripts

Gene Symbol
Ppp1r37
Entrez Gene ID
308398
Gene Name
protein phosphatase 1, regulatory subunit 37
Gene Aliases
Lrrc68
Chromosome Location
Chr. 1: 80439718 - 80471527 on Build Rnor_6.0
UniGene ID
Rn.7032
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107482.1 NP_001100952.1 4 808
GenBank mRNA BC166755.1 - 4 618

View Full Product Details