Gene Transcripts

Gene Symbol
Ankrd13d
Entrez Gene ID
361699
Gene Name
ankyrin repeat domain 13 family, member D
Gene Aliases
RGD1309156
Chromosome Location
Chr. 1: 219520452 - 219532708 on Build Rnor_6.0
UniGene ID
Rn.15704
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108514.1 NP_001101984.1 15 1769
XM_006230842.2 XP_006230904.1 15 1796
GenBank mRNA FQ213072.1 - 10 1039

View Full Product Details