Gene Transcripts

Gene Symbol
Hoxb13
Entrez Gene ID
303480
Gene Name
homeo box B13
Gene Aliases
Chromosome Location
Chr. 10: 84031955 - 84034228 on Build Rnor_6.0
UniGene ID
Rn.83970
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107041.1 NP_001100511.1 1 445

View Full Product Details