Gene Transcripts

Gene Symbol
Celf5
Entrez Gene ID
314647
Gene Name
CUGBP, Elav-like family member 5
Gene Aliases
Brunol5, RGD1565016
Chromosome Location
Chr. 7: 11095468 - 11119941 on Build Rnor_6.0
UniGene ID
Rn.12319
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135603.1 NP_001129075.1 8 985
XM_006240941.2 XP_006241003.2 8 979
XM_006240942.2 XP_006241004.2 8 952
XM_008765104.1 XP_008763326.1 7 788
XM_008765105.1 XP_008763327.1 8 977
XM_008765106.1 XP_008763328.1 8 979

View Full Product Details