Gene Transcripts

Gene Symbol
Tpbpa
Entrez Gene ID
64509
Gene Name
trophoblast specific protein alpha
Gene Aliases
Tpbp
Chromosome Location
Chr. 17: 4165041 - 4167411 on Build Rnor_6.0
UniGene ID
Rn.8816
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172073.1 NP_742070.1 2 146
GenBank mRNA AB009890.1 - 2 146

View Full Product Details