Gene Transcripts

Gene Symbol
Avp
Entrez Gene ID
24221
Gene Name
arginine vasopressin
Gene Aliases
DI, VP, Vas
Chromosome Location
Chr. 3: 123117482 - 123119460 on Build Rnor_6.0
UniGene ID
Rn.9976
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016992.2 NP_058688.2 1 92
GenBank mRNA K01495.1 - 1 73
K02434.1 - 1 73
M25646.1 - 1 92

View Full Product Details