Gene Transcripts

Gene Symbol
Klk1c7
Entrez Gene ID
24523
Gene Name
kallikrein 1-related peptidase C7
Gene Aliases
KALA, Kal, Klk1, Klk1l, Klk5, Klk7, RATKALA, RGK-7, RSKG-7
Chromosome Location
Chr. 1: 100131562 - 100135556 on Build Rnor_6.0
UniGene ID
Rn.207160
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012593.1 NP_036725.1 1 15

View Full Product Details