Gene Transcripts

Gene Symbol
Npw
Entrez Gene ID
259224
Gene Name
neuropeptide W
Gene Aliases
Chromosome Location
Chr. 10: 14021187 - 14022452 on Build Rnor_6.0
UniGene ID
Rn.14575
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153294.1 NP_695206.1 2 618
GenBank mRNA AB084278.1 - 2 618

View Full Product Details