Gene Transcripts

Gene Symbol
Stab2
Entrez Gene ID
282580
Gene Name
stabilin 2
Gene Aliases
FEEL-2, Hare
Chromosome Location
Chr. 7: 27369853 - 27552078 on Build Rnor_6.0
UniGene ID
Rn.127827
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001246357.1 NP_001233286.1 3529
GenBank mRNA AY007370.1 - 2 163
GQ477370.2 - 3529

View Full Product Details