Gene Transcripts

Gene Symbol
Fibp
Entrez Gene ID
282837
Gene Name
fibroblast growth factor (acidic) intracellular binding protein
Gene Aliases
Chromosome Location
Chr. 1: 220840083 - 220844405 on Build Rnor_6.0
UniGene ID
Rn.18229
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172334.2 NP_758837.1 9 995
GenBank mRNA AY093421.1 - 9 996

View Full Product Details