Gene Transcripts

Gene Symbol
Shcbp1l
Entrez Gene ID
289101
Gene Name
SHC SH2-domain binding protein 1-like
Gene Aliases
RGD1305609
Chromosome Location
Chr. 13: 70852044 - 70882336 on Build Rnor_6.0
UniGene ID
Rn.198417
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105961.2 NP_001099431.1 8 1381
GenBank mRNA BC160910.1 - 8 1381
FQ220197.1 - 3 550
FQ229695.1 - 3 550

View Full Product Details