Gene Transcripts

Gene Symbol
Tma16
Entrez Gene ID
290686
Gene Name
translation machinery associated 16 homolog (S. cerevisiae)
Gene Aliases
RGD1305222
Chromosome Location
Chr. 16: 24978431 - 24997286 on Build Rnor_6.0
UniGene ID
Rn.27406
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169102.1 NP_001162573.1 2 133
XM_006253023.2 XP_006253085.1 2 229
GenBank mRNA CF109688.1 - 2 168

View Full Product Details