Gene Transcripts

Gene Symbol
Ifitm1
Entrez Gene ID
293618
Gene Name
interferon induced transmembrane protein 1
Gene Aliases
Chromosome Location
Chr. 1: 213765831 - 213767841 on Build Rnor_6.0
UniGene ID
Rn.22087
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106314.1 NP_001099784.1 2 240
XM_006230530.1 XP_006230592.1 2 323

View Full Product Details