Gene Transcripts

Gene Symbol
Ms4a7
Entrez Gene ID
293744
Gene Name
membrane-spanning 4-domains, subfamily A, member 7
Gene Aliases
Chromosome Location
Chr. 1: 227490777 - 227506816 on Build Rnor_6.0
UniGene ID
Rn.44037
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106338.1 NP_001099808.1 2 501
XM_008760256.1 XP_008758478.1 2 350

View Full Product Details