Gene Transcripts

Gene Symbol
Krtcap2
Entrez Gene ID
295243
Gene Name
keratinocyte associated protein 2
Gene Aliases
Chromosome Location
Chr. 2: 188561548 - 188565544 on Build Rnor_6.0
UniGene ID
Rn.103147
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106444.1 NP_001099914.1 5 895
GenBank mRNA BC167108.1 - 5 456

View Full Product Details