Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008513.1 | NP_001008513.1 | 1 | 81 | |
GenBank mRNA | BC086571.1 | - | 1 | 81 | |
FQ218040.1 | - | 1 | 105 | ||
FQ220091.1 | - | 1 | 105 | ||
FQ223815.1 | - | 1 | 107 | ||
FQ225478.1 | - | 1 | 105 | ||
FQ226079.1 | - | 1 | 103 | ||
FQ233622.1 | - | 1 | 105 |