Gene Transcripts

Gene Symbol
Cela3b
Entrez Gene ID
298567
Gene Name
chymotrypsin-like elastase family, member 3B
Gene Aliases
Ela3b
Chromosome Location
Chr. 5: 155763877 - 155772048 on Build Rnor_6.0
UniGene ID
Rn.11639
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106692.1 NP_001100162.1 7 764
XM_008764220.1 XP_008762442.1 7 814
GenBank mRNA FQ231865.1 - 7 744

View Full Product Details