Gene Transcripts

Gene Symbol
Pramef5
Entrez Gene ID
298613
Gene Name
PRAME family member 5
Gene Aliases
Chromosome Location
Chr. 5: 98998197 - 99000744 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006238356.2 XP_006238418.1 3 1381

View Full Product Details