Gene Transcripts

Gene Symbol
Ptrhd1
Entrez Gene ID
298861
Gene Name
peptidyl-tRNA hydrolase domain containing 1
Gene Aliases
RGD1561792
Chromosome Location
Chr. 6: 28663602 - 28666987 on Build Rnor_6.0
UniGene ID
Rn.8319
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134524.1 NP_001127996.1 2 351

View Full Product Details