Gene Transcripts

Gene Symbol
Slurp1
Entrez Gene ID
300016
Gene Name
secreted Ly6/Plaur domain containing 1
Gene Aliases
RGD1308768
Chromosome Location
Chr. 7: 115961655 - 115963071 on Build Rnor_6.0
UniGene ID
Rn.20329
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130544.1 NP_001124016.1 3 331

View Full Product Details