Gene Transcripts

Gene Symbol
Lypd2
Entrez Gene ID
300017
Gene Name
Ly6/Plaur domain containing 2
Gene Aliases
RGD1307892
Chromosome Location
Chr. 7: 115967267 - 115969492 on Build Rnor_6.0
UniGene ID
Rn.19695
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130545.1 NP_001124017.1 3 418
XM_006241726.2 XP_006241788.1 3 606

View Full Product Details