Gene Transcripts

Gene Symbol
Klhdc7b
Entrez Gene ID
300150
Gene Name
kelch domain containing 7B
Gene Aliases
RGD1562257
Chromosome Location
Chr. 7: 130357643 - 130359448 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_235563.5 XP_235563.5 1 1298

View Full Product Details