Gene Transcripts

Gene Symbol
Kctd17
Entrez Gene ID
300317
Gene Name
potassium channel tetramerization domain containing 17
Gene Aliases
RGD1311154
Chromosome Location
Chr. 7: 119647229 - 119677405 on Build Rnor_6.0
UniGene ID
Rn.198231
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008765708.1 XP_008763930.1 6, 7 912
XM_008765709.1 XP_008763931.1 6, 7 910

View Full Product Details