Gene Transcripts

Gene Symbol
Zcchc13
Entrez Gene ID
302399
Gene Name
zinc finger, CCHC domain containing 13
Gene Aliases
Chromosome Location
Chr. X: 74497112 - 74498299 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003752085.3 XP_003752133.1 1 369

View Full Product Details