Gene Transcripts

Gene Symbol
Prss22
Entrez Gene ID
302971
Gene Name
protease, serine, 22
Gene Aliases
Chromosome Location
Chr. 10: 13370098 - 13374808 on Build Rnor_6.0
UniGene ID
Rn.137172
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106984.1 NP_001100454.1 6 907
GenBank mRNA AJ005642.1 - 5 891

View Full Product Details