Gene Transcripts

Gene Symbol
Ybx2
Entrez Gene ID
303250
Gene Name
Y box binding protein 2
Gene Aliases
RGD1305068
Chromosome Location
Chr. 10: 56546689 - 56552338 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_220618.8 XP_220618.3 5 742

View Full Product Details