Gene Transcripts

Gene Symbol
Rapgefl1
Entrez Gene ID
303515
Gene Name
Rap guanine nucleotide exchange factor (GEF)-like 1
Gene Aliases
Chromosome Location
Chr. 10: 86749217 - 86765678 on Build Rnor_6.0
UniGene ID
Rn.17949
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008768202.1 XP_008766424.1 1 843
XM_008768203.1 XP_008766425.1 1 844

View Full Product Details