Gene Transcripts

Gene Symbol
Arsi
Entrez Gene ID
307404
Gene Name
arylsulfatase family, member I
Gene Aliases
RGD1310242
Chromosome Location
Chr. 18: 56180089 - 56186068 on Build Rnor_6.0
UniGene ID
Rn.202490
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047881.1 NP_001041346.1 2 519

View Full Product Details