Gene Transcripts

Gene Symbol
Nfkbid
Entrez Gene ID
308496
Gene Name
nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, delta
Gene Aliases
RGD1308055, Ta-nfkbh
Chromosome Location
Chr. 1: 88883817 - 88892543 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748825.3 XP_003748873.1 4 708
XM_006228804.2 XP_006228866.1 3 1194

View Full Product Details