Gene Transcripts

Gene Symbol
Txlnb
Entrez Gene ID
308622
Gene Name
taxilin beta
Gene Aliases
RGD1307979
Chromosome Location
Chr. 1: 12915781 - 12961680 on Build Rnor_6.0
UniGene ID
Rn.165855
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135859.1 NP_001129331.1 10 1802
XM_006227657.2 XP_006227719.1 10 1787

View Full Product Details