Gene Transcripts

Gene Symbol
Setdb1
Entrez Gene ID
689883
Gene Name
SET domain, bifurcated 1
Gene Aliases
Chromosome Location
Chr. 2: 196495867 - 196527412 on Build Rnor_6.0
UniGene ID
Rn.140824
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271175.1 NP_001258104.1 18 3364
XM_006232913.2 XP_006232975.1 19 3754
XM_006232914.2 XP_006232976.1 18 3535
XM_006232916.2 XP_006232978.1 14 2731
XM_006232917.2 XP_006232979.1 14 3234

View Full Product Details