Gene Transcripts

Gene Symbol
Emr1
Entrez Gene ID
316137
Gene Name
EGF-like module containing, mucin-like, hormone receptor-like 1
Gene Aliases
Chromosome Location
Chr. 9: 9431888 - 9585875 on Build Rnor_6.0
UniGene ID
Rn.205884
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007557.1 NP_001007558.1 4 309
XM_006244280.2 XP_006244342.1 4 330
XM_008766759.1 XP_008764981.1 3 365
GenBank mRNA AY686632.1 - 4 309

View Full Product Details