Gene Transcripts

Gene Symbol
Ube2m
Entrez Gene ID
361509
Gene Name
ubiquitin-conjugating enzyme E2M
Gene Aliases
Chromosome Location
Chr. 1: 65537640 - 65541177 on Build Rnor_6.0
UniGene ID
Rn.15135
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108471.1 NP_001101941.1 4 857
GenBank mRNA FQ209516.1 - 5 450

View Full Product Details